Learn More
ACROBiosystems Influenza A [A/Thailand/1(KAN-1)/2004 (H5N1)] Neuraminidase (NA) Protein, His Tag
Marke: ACROBiosystems H54-V5247-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Neuraminidase, on the other hand, cleaves the HA-sialic acid bondage from the newly formed virions and the host cell receptors during budding. Neuraminidase thus is described as a receptor-destroying enzyme which facilitates virus release and efficient spread of the progeny virus from cell to cell.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 46.1 kDa. The protein migrates as 50-57 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
46.1 kDa | |
Influenza A [A/Thailand/1(KAN-1)/2004 (H5N1)] NA, His Tag (H54-V5247) is expressed from human 293 cells (HEK293). It contains AA His 36 - Lys 449 (Accession # H8PF47). | |
-20°C | |
Neuraminidase (NA) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
100 μg | |
His 36 - Lys 449 | |
NA,Neuraminidase | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |