Learn More
ACROBiosystems Human LAMP1 / CD107a Protein, His Tag
Marke: ACROBiosystems LA1-H5221-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Lysosomal-associated membrane protein 1 (LAMP1) is also known as CD107a (Cluster of Differentiation 107a). LAMP1 / CD107a belongs to the LAMP family. This glycoprotein provides selectins with carbohydrate ligands. It may also play a role in tumor cell metastasis. CD107a has also been shown to be a marker of degranulation on lymphocytes such as CD8+ and NK cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 55-95 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
NP_005552 | |
100 μg | |
Ala 29 - Met 382 | |
LAMP1,CD107a | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.2 kDa | |
Human LAMP1, His Tag (LA1-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Met 382 (Accession # NP_005552). | |
-20°C | |
LAMP1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |