Learn More
ACROBiosystems Human BCMA / TNFRSF17 Protein,His Tag DMF Filed
HPLC-verified
Marke: ACROBiosystems BCA-H522Y-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 12 kDa and 14-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001183.2 | |
100 μg | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
7.8 kDa | |
Human BCMA Protein, His Tag (HPLC-verified) (BCA-H522y) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). | |
-20°C | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |