Learn More
ACROBiosystems Mouse IgG1 Fc Protein, Tag Free
Marke: ACROBiosystems IG1-M5208-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.
This protein carries no "e;tag"e;. The protein has a calculated MW of 24.3 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAK53870.1 | |
200 μg | |
Val 111 - Lys 324 | |
IgG1 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.3 kDa | |
Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession # AAK53870.1). | |
-20°C | |
IgG1 Fc | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |