Learn More
ACROBiosystems Human FABP6 Protein, His Tag
Marke: ACROBiosystems FA6-H5141-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Fatty acid-binding protein 6 (FABP6), is also known as Intestinal bile acid-binding protein (I-BABP), Ileal lipid-binding protein (ILBP), Intestinal 15 kDa protein (I-15P), Gastrotropin (GT). FABP6 is a cytoplasm protein which belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. Isoform 2 of FABP6 is expressed in colorectal adenocarcinomas and their adjacent normal mucosa (at protein level). Isoform 1 of FABP6 is expressed in the jejunum, ileum, cecum and ascending colon intestine. Ileal protein FABP6 stimulates gastric acid and pepsinogen secretion. FABP6 seems to be able to bind to bile salts and bilirubins. Isoform 2 of FABP6 is essential for the survival of colon cancer cells to bile acid-induced apoptosis.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH22489 | |
100 μg | |
Ala 2 - Ala 128 | |
FABP6,ILBP,ILLBP,Gastrotropin,GT,I-15 P,I-BABP,I-BALB,I-BAP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
15.3 kDa | |
Human FABP6, His Tag (FA6-H5141) is expressed from E.coli cells. It contains AA Ala 2 - Ala 128 (Accession # AAH22489). | |
-20°C | |
FABP6 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |