Learn More
ACROBiosystems Biotinylated Human LILRA1 / CD85i / LIR-6 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems LI1-H82E8-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Leukocyte immunoglobulin-like receptors (LILRs) are inhibitory, stimulatory or soluble receptors encoded within the leukocyte receptor complex. Some LILRs are extensively polymorphic, and exhibit evidence for balancing selection and association with disease susceptibility. LILRA1 (LIR-6) can recognize MHC (major histocompatibility complex) class I or class I-like molecules, and high levels of sequence similarity among LILRA1, LILRA2 (ILT1), LILRA3 (ILT6) and LILRB1/B2.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 52.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP 006854.1 | |
25 μg | |
Pro 17 - Asn 461 | |
LILRA1,LIR6,CD85i,LIR-6 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
52.2 kDa | |
Biotinylated Human LILRA1, His,Avitag (LI1-H82E8) is expressed from human 293 cells (HEK293). It contains AA Pro 17 - Asn 461 (Accession # O75019-1). | |
-20°C | |
LILRA1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |