Learn More
ACROBiosystems Human Syndecan-1 Protein, His Tag
Marke: ACROBiosystems SD1-H5228-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Syndecan-1 (SYND1 or SDC1) is also known as CD antigen CD138, is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-1 / SDC1 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. It is a useful marker for plasma cells, but only if the cells tested are already known to be derived from blood.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 44 kDa and 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH08765 | |
100 µg | |
Gln 23 - Gly 254 | |
SDC1,Syndecan-1,CD138,SYND1,SDC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.7 kDa | |
Human Syndecan-1, His Tag (SD1-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # AAH08765). | |
-20°C | |
Syndecan-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |