Learn More
ACROBiosystems Human SIRP beta / CD172b Protein, Fc Tag
Marke: ACROBiosystems SIA-H5257-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Signal-regulatory protein beta-1 (SIRP beta 1) also known as SIRPβ1, belongs to signal-regulatory-protein (SIRP) family, and immunoglobulin superfamily. SIRPβ1 is a transmembrane protein that has three Ig-like domains in its extracellular region and a short cytoplasmic tail. Human SIRPβ1 is expressed on monocytes and granulocytes but not on lymphocytes. SIRP beta 1 contains short cytoplasmic domains that lack cytoplasmic sequence motifs capable of recruiting SHP-2 and SHP-1. In addition, they contain a single basic lysine residue within the hydrophobic transmembrane domain.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006056.2 | |
100 μg | |
Glu 30 - Leu 371 | |
CD172b antigen,CD172b,DKFZp686A05192,FLJ26614,SIRP beta 1,SIRPB1,SIRP-BETA-1,9930027N05Rik,SIRP-beta | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
63.7 kDa | |
Human SIRP beta, Fc Tag (SIA-H5257) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Leu 371 (Accession # O00241-1). | |
-20°C | |
SIRP beta | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |