Learn More
ACROBiosystems Biotinylated Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein, His, Avitag™&Tag Free
Biotinylated Protein
Marke: ACROBiosystems IT1-H82Wb-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Human Integrin alpha 8 beta 1 Heterodimer Protein consists of ITGA8 and ITGB1. The integrin alpha 8 subunit, isolated by low stringency hybridization, is a novel integrin subunit that associates with beta 1. The recently identified alpha 8 integrin subunit associates with beta 1 and is predominantly expressed in smooth muscle and other contractile cells in adult tissues, and in mesenchymal and neural cells during development. In addition, Integrin alpha 8 beta 1 is a receptor for fibronectin and can promote attachment, cell spreading, and neurite outgrowth on fibronectin.
Biotinylated Human ITGA8&ITGB1 Heterodimer Protein, His,Avitag&Tag Free, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 116.4 kDa (ITGA8) and 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followedSpezifikation
NP 003629.2/NP_002202.2 | |
25 μg | |
Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) | |
Integrin alpha 8 beta 1,ITGA8andITGB1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
116.4 kDa (ITGA8) and 83.7 kDa (ITGB1) | |
Biotinylated Human ITGA8&ITGB1; Heterodimer Protein, His,AviTag&Tag; Free (IT1-H82Wb) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) and P05556-1 (ITGB1)). | |
-20°C | |
Integrin alpha 8 beta 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |