Learn More
ACROBiosystems Human ICOS / CD278 Protein, Fc Tag
Marke: ACROBiosystems ICS-H5258-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Inducible T-cell costimulator (ICOS) is also known as Activation-inducible lymphocyte immunomediatory molecule (AILIM), CD278, which belongs to the CD28 family of immune costimulatory receptors consisting of CD28, CTLA-4 and PD-1. ICOS enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells. CD278 / ICOS prevents the apoptosis of pre-activated T-cells and also plays a critical role in CD40-mediated class switching of immunoglobin isotypes.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_036224.1 | |
100 μg | |
Glu 21 - Phe 141 | |
ICOS,CD278,AILIM,Inducible T-cell costimulator | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
40.2 kDa | |
Human ICOS, Fc Tag (ICS-H5258) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Phe 141 (Accession # Q9Y6W8-1). | |
-20°C | |
ICOS | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |