Learn More
ACROBiosystems Human IL-13 Protein, Fc Tag
Marke: ACROBiosystems IL3-H5256-500ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interleukin 13 (IL13) is also known as ALRH, BHR1and P600, is a single-chain glycosylated polypeptide, and is a cytokine critical in regulating inflammatory and immune responses. IL13 is secreted by many cell types, but especially by T helper type 2 (Th2) cells. IL-13 induces its effects through a multi-subunit receptor that includes the alpha chain of the IL-4 receptor (IL-4Rα) and at least one of two known IL-13-specific binding chains. The functions of IL-13 overlap considerably with those of IL-4, especially with regard to changes induced on hematopoietic cells, but these effects are probably less important given the more potent role of IL-4. IL-13 induces matrix metalloproteinases (MMPs) as part of a mechanism that protects against excessive allergic inflammation that predisposes to asphyxiation. IL-13 induces many features of allergic lung disease, including airway hyperresponsiveness, goblet cell metaplasia and mucus hypersecretion, which all contribute to airway obstruction.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAK53823.1 | |
500 μg | |
Gly 21 - Asn 132 | |
IL13,ALRH,BHR1,MGC116786,MGC116788,MGC116789,P600,Interleukin-13 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
38.8 kDa | |
Human IL-13, Fc Tag (IL3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1). | |
-20°C | |
IL-13 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |