Learn More
ACROBiosystems Human Thrombospondin-2 Protein, His Tag
Marke: ACROBiosystems TH2-H52H5-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Thrombospondin-2 is also known as THBS2, TSP2, TSP-2. Thrombospondin-2 (TSP2) is an anti-angiogenic matricellular protein that inhibits tumor growth and angiogenesis. Adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. Ligand for CD36 mediating antiangiogenic properties. 2 SNPs in the intron region of THBS2 gene (rs6422747 and rs6422748) were associated with susceptibility of IDD. However, they were not related with severity of IDD, including the total number of degenerative disc and level of IDD. Thrombospondin 2, an important regulator of synaptogenesis in central nervous system development, was distributed in retinal macroglia cells, and its receptor α2δ-1 was in retinal neurons. activated macroglia cells may participate in alterations of presynaptic proteins of retinal neurons following elevated hydrostatic pressure, and macroglia-derived thrombospondin 2 may modulate these changes via binding to its neuronal receptor α2δ-1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 129.7 kDa. The protein migrates as 160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003238.2 | |
100 μg | |
Gly 19 - Ile 1172 | |
Thrombospondin-2,THBS2,TSP2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
129.7 kDa | |
Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession # P35442-1). | |
-20°C | |
Thrombospondin-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |