Learn More
ACROBiosystems Human Apolipoprotein A-II / APOA2 Protein, His Tag
Marke: ACROBiosystems APA-H51H6-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Apolipoprotein A-II(APO-A2) stabilize HDL (high density lipoprotein) structure by its association with lipids, and affect the HDL metabolism. It is produced by the liver and is involved in cholesteryl ester formation and cholesterol transport from tissues to the liver. Recent studies have established the interaction between APOA2 -256T>C polymorphism and dietary saturated fatty acids intake in relation to obesity on healthy individuals. Significant interaction between APOA2-saturated fatty acids intake and body mass index (BMI). The result of this study confirmed the interaction between APOA2 -256T>C polymorphism and SFAs intake with BMI in type 2 diabetic patients.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.3 kDa. The protein migrates as 11 kDa and 13 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001634.1 | |
100 μg | |
Ala 19 - Gln 100 | |
Apolipoprotein A-II,APOA2,Apo-AII,ApoA-II,Apolipoprotein A2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
11.3 kDa | |
Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed from E.coli cells. It contains AA Ala 19 - Gln 100 (Accession # P02652-1). | |
-20°C | |
Apolipoprotein A-II | |
Human | |
PBS, pH7.4 | |
E.coli | |
Other Recombinant Proteins |