Learn More
ACROBiosystems Human CD30 Ligand / TNFSF8 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems CDL-H525b-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD30 ligand (CD30L) and CD30 are interacting cellsurface glycoproteins that are members of the tumour necrosis factor (TNF) and tumour necrosis factor receptor (TNFR) superfamilies. In humans, CD30L is expressed at high levels on activated T cells. There is conflicting evidence with regard to cell surface expression of CD30L on primary human B cells. An initial report of constitutive CD30L expression on peripheral blood B cells33 has not been substantiated.
This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001235.1 | |
100 μg | |
Gln 63 - Asp 234 | |
CD30-L,CD153,TNFSF8,CD30L,CD30LG,CD153 antigen,CD30 antigen ligand,CD30 Ligand | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
46.5 kDa | |
Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H525b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (Accession # P32971-1). | |
-20°C | |
CD30 ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |