Learn More
ACROBiosystems Biotinylated Human OX40 Ligand / TNFSF4 Protein, Avitag™,Fc Tag
Biotinylated Protein
Marke: ACROBiosystems OXL-H82F4-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor ligand superfamily member 4 (TNFSF4) is also known as glycoprotein Gp34, OX40 ligand (OX40L), TAX transcriptionally-activated glycoprotein 1 and CD252, which belongs to the tumor necrosis factor family. TNFSF4 is the ligand for CD134 and is expressed on such cells as DC2s (a subtype of dendritic cells) enabling amplification of Th2 cell differentiation. The interaction of TNFSF4-TNFSF4 is involved in the pathogenesis of multiple autoimmune and inflammatory diseases such as systemic lupus erythematosus (SLE), carotid artery disease and cancer. Furthermore, similar to other TNF superfamily members, membrane-bound OX40 Ligand (TNFSF4) exists as a homotrimer. Human TNFSF4 shares 46% amino acid sequence identity with its mouse counterpart.
This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 48.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003317.1 | |
25 μg | |
Gln 51 - Leu 183 | |
OX40L,TNFSF4,CD252,Glycoprotein Gp34,TXGP1,CD134 ligand,CD134L | |
Precision Avi | |
Avi Tag and Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.7 kDa | |
Biotinylated Human O x 40 Ligand, Avitag,Fc Tag (OXL-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1). | |
-20°C | |
OX40 Ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |