Learn More
ACROBiosystems Biotinylated Human HE4 / WFDC2 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Human Epididymis Protein 4 (HE4) is also known as WAP four-disulfide core domain protein 2 (WFDC2), Epididymal secretory protein E4, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, is a member of the WFDC domain family. HE4 / WFDC2 is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. HE4 is also a small secretory protein, which may be involved in sperm maturation.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 13.7 kDa. The protein migrates as 19-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006094.3 | |
200 μg | |
Glu 31 - Phe 124 | |
HE4,WAP5,WFDC2,EDDM4 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
13.7 kDa | |
Biotinylated Human HE4, His,Avitag (HE4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Phe 124 (Accession # Q14508-1). | |
-20°C | |
HE4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |