Learn More
ACROBiosystems Human MAG / Siglec-4a Protein, His Tag
Marke: ACROBiosystems MAG-H52H8-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Myelin-associated glycoprotein (MAG), a nervous system cell adhesion molecule, is an I-type lectin that binds to sialylated glycoconjugates, including gangliosides bearing characteristic structural determinants. Preferentially binds to alpha-2,3-linked sialic acid. Binds ganglioside Gt1b. Adhesion molecule that mediates interactions between myelinating cells and neurons by binding to neuronal sialic acid-containing gangliosides and to the glycoproteins RTN4R and RTN4RL2. Protection against apoptosis is probably mediated via interaction with neuronal RTN4R and RTN4RL2. In dorsal root ganglion neurons the inhibition is mediated primarily via binding to neuronal RTN4R or RTN4RL2 and to a lesser degree via binding to neuronal gangliosides. In cerebellar granule cells the inhibition is mediated primarily via binding to neuronal gangliosides. In sensory neurons, inhibition of neurite extension depends only partially on RTN4R, RTN4RL2 and gangliosides.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002352.1 | |
100 μg | |
Gly 20 - Pro 516 | |
MAG,Siglec-4 A,GMA,S-MAG | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
56.6 kDa | |
Human MAG, His Tag (MAG-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1). | |
-20°C | |
MAG / Siglec-4 A | |
Human | |
PBS with Arginine, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |