Learn More
ACROBiosystems Biotinylated Human EpCAM / TROP1 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems EPM-H82F9-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
EpCAM is also known as CO171A, EGP, EGP40,GA7332, KSA, M4S, MIC18, MK1, TROP1, hEGP2, and is a pan-epithelial differentiation antigen that is expressed on almost all carcinomas as 17-1A(mAb) antigen. Its constitutional function is being elucidated. It is intricately linked with the Cadherin-Catenin pathway and hence the fundamental WNT pathway responsible for intracellular signaling and polarity. The epithelial cell adhesion molecule (Ep-CAM) is known to express in most epithelial malignancies and was reported as a tumor marker or a candidate of molecular targeting therapy. Ep-CAM cross signaling with N-cadherin involves Pi3K, resulting in the abrogation of the cadherin adhesion complexes in epithelial cells was reported. And Epithelial cell adhesion molecule (Ep-CAM) recently received increased attention as a prognostic factor in breast cancer.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 56.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH14785.1 | |
25 μg | |
Gln 24 - Lys 265 | |
EPCAM,TACSTD1,TROP1,CD326,DIAR5,EGP2,EGP314,EGP40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MK1 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
56.2 kDa | |
Biotinylated Human EpCAM, Fc,Avitag (EPM-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). | |
-20°C | |
EpCAM | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |