Learn More
ACROBiosystems PE-Labeled Human PD-1 / PDCD1 Protein, Fc Tag, His Tag (recommended for neutralizing assay)
Marke: ACROBiosystems PD1-HP2F2-250tests
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 44.7 kDa.Spezifikation
NP_005009.2 | |
250 tests | |
Leu 25 - Gln 167 | |
PDCD1,PD1,CD279,SLEB2 | |
PE Labeled | |
His Tag and Fc Tag | |
HEK293 | |
Fluorescent labeled protein |
44.7 kDa | |
PE-Labeled Human PD-1, Fc tag, His tag (recommended for neutralizing assay) (Cat. No. PD1-HP2F2) is produced via conjugation of PE to Human PD-1, Fc tag, His tag under optimal conditions. Human PD-1, Fc tag, His tag is expressed in HEK293 cells containing AA Leu 25 - Gln 167 (Accession # Q15116-1). | |
-20°C, avoid light | |
PD-1 | |
Human | |
PBS, 0.5% BSA, pH7.4 | |
Powder |