Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag
Marke: ACROBiosystems NUN-C51H9-100UG
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 49.4 kDa. The protein migrates as 47-50 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
QHO62115.1 | |
100 μg | |
Met 1 - Ala 419 | |
Nucleocapsid protein,NP,Protein N,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
49.4 kDa | |
SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C51H9) is expressed from E.coli cells. It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). | |
-20°C | |
Nucleocapsid protein | |
SARS-CoV-2 | |
50 mM Tris, 150 mM NaCl, Arginine, pH7.5 | |
E.coli | |
Viral Proteins |