Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S2 protein, His Tag
Marke: ACROBiosystems S2N-C52H5-100UG
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
QHD43416.1 | |
100 μg | |
Ser 686 - Pro 1213 | |
Spike,S2 protein,Spike glycoprotein Subunit2,S glycoprotein Subunit2,Spike protein S2,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
60.0 kDa | |
SARS-CoV-2 S2 protein, His Tag (S2N-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to prevent the formation of aggregates in the course of protein production. | |
-20°C | |
S2 protein | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |