Learn More
ACROBiosystems Biotinylated SARS-CoV-2 (COVID-19) S protein RBD, His,Avitag™
MALS verified
Marke: ACROBiosystems SPD-C82E9-25UG
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa. The protein migrates as 33-36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
QHD43416.1 | |
25 μg | |
Arg 319 - Phe 541 | |
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
28.2 kDa | |
Biotinylated SARS-CoV-2 S protein RBD, His,Avitag (SPD-C82E9) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1). | |
-20°C | |
S protein RBD | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |