Learn More
ACROBiosystems HCoV-HKU1(isolate N5) S1 protein, His Tag
Marke: ACROBiosystems SIN-V52H6-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.7 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Spezifikation
85.7 kDa | |
HCoV-HKU1(isolate N5) S1 protein, His Tag (SIN-V52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 13 - Arg 756 (Accession # Q0ZME7-1). | |
-20°C | |
S1 protein | |
HCoV-HKU1 (isolate N5) | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
100 μg | |
Ala 13 - Arg 756 | |
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |