missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike NTD Neutralizing Mouse anti-SARS-CoV-2, Unconjugated, Clone: AM121, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems SPD-M121-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Spike NTD Neutralizing Monoclonal antibody specifically detects Spike NTD Neutralizing in SARS-CoV-2 samples.Spezifikation
Spike NTD Neutralizing | |
AM121 | |
Human (HEK293) | |
RUO | |
SARS-CoV-2 | |
IgG1 |
Monoclonal | |
Unconjugated | |
100 μg | |
Primary | |
-20°C |