Learn More
ACROBiosystems Human NCAM-1 / CD56 Protein, Fc Tag
MALS verified
Marke: ACROBiosystems NC1-H5253-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
NCAM1 belongs to the immunoglobulin superfamily of adhesion molecules. A wide range of alternatively spliced NCAM1 messenger RNAs (mRNAs) has been described to date, but only the 120-, 140-, and 180- kDa isoforms are commonly expressed. NCAM1 plays an important role in the regulation of neurogenesis, neurite outgrowth, proliferation, and cell migration, however, its function in hematopoiesis, including NK cells, is poorly understood. NCAM1 signaling is mediated either by homophilic or heterophilic interactions with fibroblast growth factor receptor (FGFR), L1-CAM, N-cadherin and other components of the extracellular matrix. Upon activation, NCAM1 triggers a variety of signaling cascades including FYN–focal adhesion kinase (FAK), MAPK, and phosphatidylinositol 3-kinase (PI3K) pathways.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_851996.2 | |
100 μg | |
Leu 20 - Gly 718 | |
CD56,MSK39,NCAM1,N-CAM-1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
104.0 kDa | |
Human NCAM-1, Fc Tag (NC1-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2). | |
-20°C | |
NCAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |