Learn More
ACROBiosystems Biotinylated Human CLEC12A / MICL / CLL-1 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems CLA-H82F7-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CLEC12A (C-type lectin domain family 12 member A) is also known as CLL1, DCAL2, MICL. Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death. Cell surface receptor that modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases. Evidence of distinct disease propagating stem cells in myelodysplastic syndrome (MDS) has emerged in recent years. The role of CLEC12A in MDS, however, remains to be elucidated. Furthermore, CLEC12A has been proposed as a promising marker of leukaemic stem cells in AML.
This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 51.9 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_612210.4 | |
200 μg | |
His 65 - Ala 265 | |
CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371 | |
Precision Avi | |
Avi Tag and Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
51.9 kDa | |
Biotinylated Human CLEC12A, Fc,Avitag (CLA-H82F7) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2). | |
-20°C | |
CLEC12A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |