missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Oxazol-5-yl)-anilin, ≥ 97 %, Thermo Scientific™
Click to view available options
Menge:
1 g
5 g
Spezifikation
Chemischer Name oder Material | 2-(1, 3-Oxazol-5-yl)aniline |
CAS Min. % | 89.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 90% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 1 g |
Synonym | 2-1,3-oxazol-5-yl aniline, 2-5-oxazolyl aniline, 2-oxazol-5-yl aniline, benzenamine, 2-5-oxazolyl, 2-oxazol-5-yl phenylamine, benzenamine,2-5-oxazolyl, 5-2-aminophenyl-1,3-oxazole, 2-1,3-oxazol-5-yl phenylamine |
Formelmasse | 160.18 |
Reinheit (%) | 90% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'