missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,6,8,8-Octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 98%, BLD Pharm™

Product Code. 31007356
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31007356 5 g
31000107 500 g
30993832 100 g
30989382 1 kg
30990190 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 31007356 missing translation for 'mfr' BLD Pharm missing translation for 'supplierNo' BD1460765g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 2,2,4,4,6,6,8,8-Octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
CAS 546-56-5
Summenformel C48H40O4Si4
MDL-Nummer MFCD00003268
Menge 5 g
SMILES C1([Si]2(O[Si](C3=CC=CC=C3)(O[Si](C4=CC=CC=C4)(O[Si](C5=CC=CC=C5)(O2)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)=CC=CC=C1
Molekulargewicht (g/mol) 793.17
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.