missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichlorophenyl)acetonitrile, 98%, BLD Pharm™

Product Code. 30968067
Encompass_Preferred
Change view
Click to view available options
Menge:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30968067 25 g
30952751 5 g
30964805 100 g
30955571 10 g
30958000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30968067 Supplier BLD Pharm Supplier No. BD1088125g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 2-(2,6-Dichlorophenyl)acetonitrile
CAS 3215-64-3
Summenformel C8H5Cl2N
MDL-Nummer MFCD00001901
Menge 25 g
SMILES C1=CC=C(C(=C1Cl)CC#N)Cl
Molekulargewicht (g/mol) 186.04
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.