missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Thiocarbamoylphenyl)-benzimidazol, 97 %, Thermo Scientific™

Catalog Number 15498317
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1217863-37-0
C14H11N3S
253.323
MFCD16251582
ORFAWVSZCQRSSQ-UHFFFAOYSA-N
2-3-thiocarbamoylphenyl benzimidazole, 3-1h-1,3-benzodiazol-2-yl benzenecarbothioamide, 3-1h-benzo d imidazol-2-yl benzothioamide
57495752
3-(1H-Benzimidazol-2-yl)Benzolcarbothioamid
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)C(=S)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1217863-37-0
C14H11N3S
253.323
MFCD16251582
ORFAWVSZCQRSSQ-UHFFFAOYSA-N
2-3-thiocarbamoylphenyl benzimidazole, 3-1h-1,3-benzodiazol-2-yl benzenecarbothioamide, 3-1h-benzo d imidazol-2-yl benzothioamide
57495752
3-(1H-Benzimidazol-2-yl)Benzolcarbothioamid
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(3-Thiocarbamoylphenyl)benzimidazole
Menge 250 mg
Formelmasse 253.32
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'