missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulfamoylbenzoic acid, 98%, BLD Pharm™

Product Code. 30942578
Encompass_Preferred
Change view
Click to view available options
Menge:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30942578 25 g
30937269 100 g
30941852 1 kg
30939133 10 g
30940437 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30942578 Supplier BLD Pharm Supplier No. BD1034125g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 2,4-Dichloro-5-sulfamoylbenzoic acid
CAS 2736-23-4
Summenformel C7H5Cl2NO4S
MDL-Nummer MFCD00007931
Menge 25 g
SMILES O=C(O)C1=CC(S(=O)(N)=O)=C(Cl)C=C1Cl
Molekulargewicht (g/mol) 270.09
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.