missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, 97%, BLD Pharm™

Artikelnummer. 30965784
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
100 mg
250 mg
1 g
5 g
10 g
25 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30965784 10 g
30963568 25 g
30953913 100 mg
30957537 250 mg
30959728 5 g
30953236 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30965784 Lieferant BLD Pharm Lieferanten-Nr. BD11673310g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate
CAS 35013-72-0
Summenformel C14H19N3O5S
MDL-Nummer MFCD00078531
Menge 10 g
SMILES O=C(CC1)N(OC(CCCC[C@@H]2SC[C@]([C@]2([H])N3)([H])NC3=O)=O)C1=O
Molekulargewicht (g/mol) 341.38
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.