missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxybenzamid, 97 %

Catalog Number 11338155
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3147-50-0
C7H7NO3
153.14
MFCD00017125
WFIWHFWPQQSJDD-UHFFFAOYSA-N
76602
2,6-dihydroxybenzamid
NC(=O)C1=C(O)C=CC=C1O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3147-50-0
C7H7NO3
153.14
MFCD00017125
WFIWHFWPQQSJDD-UHFFFAOYSA-N
76602
2,6-dihydroxybenzamid
NC(=O)C1=C(O)C=CC=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Dihydroxybenzamide
Schmelzpunkt ∽210°C (decomposition)
Menge 1 g
Beilstein 2937308
Synonym .gamma.-resorcylamide, benzamide, 2,6-dihydroxy, g-resorcylamide, 2,6 dihydroxybenzamide, acmc-20ao8l, 2,6-dihydroxy benzamid, benzamide,2,6-dihydroxy
Formelmasse 153.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'