Learn More
2-Amino-5-Ethyl-4-Phenylthiazol, 97 %, Thermo Scientific™
Catalog Number 15428697
Click to view available options
Menge:
1 g
5 g
34176-47-1
C11H12N2S
204.291
MFCD00736900
JPFFDVPXYBZEPP-UHFFFAOYSA-N
147560
5-Ethyl-4-Phenyl-1,3-Thiazol-2-Amin
CCC1=C(N=C(S1)N)C2=CC=CC=C2
34176-47-1
C11H12N2S
204.291
MFCD00736900
JPFFDVPXYBZEPP-UHFFFAOYSA-N
147560
5-Ethyl-4-Phenyl-1,3-Thiazol-2-Amin
CCC1=C(N=C(S1)N)C2=CC=CC=C2
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Amino-5-ethyl-4-phenylthiazole
Menge 1 g
Empfindlichkeit Air Sensitive
Synonym 2-thiazolamine, 5-ethyl-4-phenyl, 2-amino-5-ethyl-4-phenylthiazole, 2-thiazolamine,5-ethyl-4-phenyl, 5-ethyl-4-phenyl-thiazol-2-ylamine, cambridge id 7169497, 5-ethyl-4-phenyl-2-thiazolamine, 5-ethyl-4-phenylthiazol-2-amine, 4-phenyl-5-ethyl-2-thiazoleamine, 2-amino-5-ethyl-4-phenyl-1,3-thiazole, 5-ethyl-4-phenyl-1,3-thiazole-2-ylamine
Formelmasse 204.29
Reinheit (%) 97%