missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-6-chlorpyridin, 95 %, Thermo Scientific™

Catalog Number 15433577
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5140-72-7
C5H3BrClN
192.44
MFCD00181262
JWTZSVLLPKTZJP-UHFFFAOYSA-N
pyridine, 2-bromo-6-chloro, abbypharma ap-18-5384, pubchem9626, 2-chloro-6-bromopyridine, acmc-1awk4, 2-bromo-6-chloro-pyridine, 2-bromo-6-chloropyridine?, ksc493o0h, 2-bromo-6-chloropyridine
78820
2-Brom-6-Chlorpyridin
C1=CC(=NC(=C1)Br)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5140-72-7
C5H3BrClN
192.44
MFCD00181262
JWTZSVLLPKTZJP-UHFFFAOYSA-N
pyridine, 2-bromo-6-chloro, abbypharma ap-18-5384, pubchem9626, 2-chloro-6-bromopyridine, acmc-1awk4, 2-bromo-6-chloro-pyridine, 2-bromo-6-chloropyridine?, ksc493o0h, 2-bromo-6-chloropyridine
78820
2-Brom-6-Chlorpyridin
C1=CC(=NC(=C1)Br)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-6-chloropyridine
Menge 250 mg
Formelmasse 192.44
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'