missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorotritylchlorid auf Polystyrol, 1 % vernetzt, 100 bis 200 Mesh, 1.0 bis1.4 mmol/g., Thermo Scientific Chemicals
Click to view available options
Menge:
2 g
10 g
50 g
Beschreibung
2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | 2-Chlorotrityl chloride on polystyrene |
Namenshinweis | 1% cross-linked, 1.0 to 1.4 mmol/g |
Physikalische Form | Pulver |
Menge | 2 g |
Netzgröße | 100 to 200 Mesh |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'