missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-phenyl-5-pyrimidincarbonsäure, 97 %, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Menge:
250 mg
1 g
10 g
Specifications
Chemischer Name oder Material | 2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid |
CAS Min. % | 96.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 97% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 250 mg |
Synonym | 2-methyl-4-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl-4-phenyl, 5-pyrimidinecarboxylic acid, 2-methyl-4-phenyl, acmc-1c0qx, 2-methyl-4-phenylpyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid,2-methyl-4-phenyl, 2-methyl-4-phenyl-pyrimidine-5-carboxylic acid |
Formelmasse | 214.22 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'