Learn More
Human RBFOX3/NeuN Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB11665-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human samples. It is validated for Multiplex Immunofluorescence,Western Blot,Immunohistochemistry
Spezifikation
| RBFOX3/NeuN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3 | |
| Synthetic Peptide, Accession # A6NFN3 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Multiplex Immunofluorescence, Western Blot, Immunohistochemistry | |
| 1094645 | |
| Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL | |
| A6NFN3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 146713 | |
| Human | |
| Antibody | |
| IgG1 |