Learn More
DiscoverX PathHunter™ HEK 293 F2RL1 (PAR2, GPR11) β-Arrestin Cell Line
Artikelnummer. 17312799
Encompass
Change view
Click to view available options
Menge:
2 Fläschchen
Artikelnummer. Menge
17312799 2 Fläschchen
1 options
Artikelnummer. 17312799
Marke: DiscoverX 930235C1
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
The PathHunter™ HEK 293 F2RL1 (PAR2, GPR11) β-Arrestin Cell Line measures F2RL1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
The PathHunter™ F2RL1 (PAR2,GPR11) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. proteases, SLIGRL) based activation of F2RL1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged F2RL1 and EA-tagged β-Arrestin 2. Activation of F2RL1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Spezifikation
Assay-Eingabe β-Arrestin recruitment
Zelllinie 293H
Zur Verwendung mit (Anwendung) Cell Lines
Primärer Agonist SLIGRL-NH2
Menge 2 Fläschchen
Zieleintrag F2RL1 (PAR2,GPR11)
Zugriffsnummer NM_005242.4