missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(Boc-Amino)-2-Hydroxy-5-Methylhexansäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7034869
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73397-27-0
C12H23NO5
261.32
MFCD04974450
DJZCWTDKDFJARG-UHFFFAOYNA-N
2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, n-boc-2s,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2s,3s-2-hydroxy-5-methyl-3-2-methylpropan-2-yl oxycarbonylamino hexanoic acid, 2s,3s-3-t-butoxycarbonyl amino-2-hydroxy-5-methylhexaneoic acid, 2s,3s-2-hydroxy-3-tert-butyloxycarbonyl amino-5-methylhexanoic acid, 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoicacid, hexanoic acid,3-1,1-dimethylethoxy carbonyl amino-2-hydroxy-5-methyl-, 2s,3s
2762291
CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73397-27-0
C12H23NO5
261.32
MFCD04974450
DJZCWTDKDFJARG-UHFFFAOYNA-N
2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, n-boc-2s,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2s,3s-2-hydroxy-5-methyl-3-2-methylpropan-2-yl oxycarbonylamino hexanoic acid, 2s,3s-3-t-butoxycarbonyl amino-2-hydroxy-5-methylhexaneoic acid, 2s,3s-2-hydroxy-3-tert-butyloxycarbonyl amino-5-methylhexanoic acid, 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoicacid, hexanoic acid,3-1,1-dimethylethoxy carbonyl amino-2-hydroxy-5-methyl-, 2s,3s
2762291
CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
Menge 250 mg
IUPAC-Name 3-{[(tert-Butoxy)carbonyl]amino}-2-hydroxy-5-methylhexansäure
Formelmasse 261.32
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'