missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)acrylic acid, 98%, BLD Pharm™

Product Code. 30965786
Encompass_Preferred
Change view
Click to view available options
Menge:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30965786 100 g
30954927 500 g
30954167 1 kg
30963292 25 g
30957835 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30965786 Supplier BLD Pharm Supplier No. BD11651100g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 3-(3,4-Dihydroxyphenyl)acrylic acid
CAS 331-39-5
Summenformel C9H8O4
MDL-Nummer MFCD00004392
Menge 100 g
SMILES O=C(O)/C=C/C1=CC=C(O)C(O)=C1
Molekulargewicht (g/mol) 180.16
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.