Learn More
3-(3 -Ethyl-1,2,4-oxadiazol-5-yl)piperidin, 97 %, Thermo Scientific™
Catalog Number 11469570
Click to view available options
Menge:
250 mg
1 g
139269-06-0
C9H15N3O
181.24
MFCD08061044
XXIODYOSZOJCLL-UHFFFAOYNA-N
19886311
3-(3-ethyl-1,2,4-oxadiazol-5-yl)piperidine
CCC1=NOC(=N1)C1CCCNC1
139269-06-0
C9H15N3O
181.24
MFCD08061044
XXIODYOSZOJCLL-UHFFFAOYNA-N
19886311
3-(3-ethyl-1,2,4-oxadiazol-5-yl)piperidine
CCC1=NOC(=N1)C1CCCNC1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
Menge 1 g
Synonym 3-3-ethyl-1,2,4-oxadiazol-5-yl piperidine, 3-ethyl-5-piperidin-3-yl-1,2,4-oxadiazole, pubchem18373, acmc-20ae3s, 3-ethyl-5-3-piperidyl-1,2,4-oxadiazole, 3-3-ethyl-1,2,4 oxadiazol-5-yl-piperidine, piperidine,3-3-ethyl-1,2,4-oxadiazol-5-yl, piperidine, 3-3-ethyl-1,2,4-oxadiazol-5-yl
Formelmasse 181.24
Reinheit (%) 97%