missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlor-alpha-Toluensulfonylchlorid, 96 %, Thermo Scientific Chemicals

Catalog Number p-7033571
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
163295-70-3
C7H5Cl3O2S
259.53
MFCD01631928
GZMJRPMBVICCFL-UHFFFAOYSA-N
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride
4993270
(3,5-Dichlorphenyl)-methansulfonylchlorid
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
163295-70-3
C7H5Cl3O2S
259.53
MFCD01631928
GZMJRPMBVICCFL-UHFFFAOYSA-N
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride
4993270
(3,5-Dichlorphenyl)-methansulfonylchlorid
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,5-Dichloro-alpha-toluenesulfonyl chloride
Menge 250 mg
UN-Nummer UN3096
Formelmasse 259.54
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'