Learn More
3'-Benzyloxycetophenon, 97 %, Thermo Scientific Chemicals
Catalog Number 11476087
Click to view available options
Menge:
5 g
25 g
100 g
34068-01-4
C15H14O2
226.275
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene
98689
1-(3-Phenylmethoxyphenyl)ethanon
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
34068-01-4
C15H14O2
226.275
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene
98689
1-(3-Phenylmethoxyphenyl)ethanon
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3'-Benzyloxyacetophenone
Menge 5 g
Formelmasse 226.28
Reinheit (%) 97%