missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-4-(4-morpholinyl)-benzolboronsäurepinakolester, 95 %, Thermo Scientific™

Catalog Number 15484896
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1361110-63-5
C16H23BClNO3
323.62
MFCD18070979
SKJAWVMUHVUHMM-UHFFFAOYSA-N
4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-chloro-4-morpholinophenylboronic acid pinacol ester, 3-chloro-4-morpholinophenylboronicacidpinacolester, 3-chloro-4-4-morpholinyl benzeneboronic acid pinacol ester, 3-chloro-4-morpholinophenyl boronic acid pinacol ester, 4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl morpholine, 4-2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine
56965741
4-[2-Chlor-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Morpholin
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(C=C1)N1CCOCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1361110-63-5
C16H23BClNO3
323.62
MFCD18070979
SKJAWVMUHVUHMM-UHFFFAOYSA-N
4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-chloro-4-morpholinophenylboronic acid pinacol ester, 3-chloro-4-morpholinophenylboronicacidpinacolester, 3-chloro-4-4-morpholinyl benzeneboronic acid pinacol ester, 3-chloro-4-morpholinophenyl boronic acid pinacol ester, 4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl morpholine, 4-2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine
56965741
4-[2-Chlor-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Morpholin
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(C=C1)N1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-4-(4-morpholinyl)benzeneboronic acid pinacol ester
Menge 250 mg
Formelmasse 323.63
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'