missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridin-4-boronsäurepinacolester, 95 %, Thermo Scientific Chemicals

Catalog Number p-7030255
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
878194-92-4
C12H15BN2O2
230.074
MFCD09037481
LUDBQHAQFBWGNI-UHFFFAOYSA-N
3-cyanopyridine-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 3-cyano-4-pyridineboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyanopyridin-4-ylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-pyridinecarbonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarbonitrile, 3-cyanopyridine-4-boronicacidpinacolester
44755176
4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin-3-Carbonitril
B1(OC(C(O1)(C)C)(C)C)C2=C(C=NC=C2)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
878194-92-4
C12H15BN2O2
230.074
MFCD09037481
LUDBQHAQFBWGNI-UHFFFAOYSA-N
3-cyanopyridine-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 3-cyano-4-pyridineboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyanopyridin-4-ylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-pyridinecarbonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarbonitrile, 3-cyanopyridine-4-boronicacidpinacolester
44755176
4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin-3-Carbonitril
B1(OC(C(O1)(C)C)(C)C)C2=C(C=NC=C2)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Cyanopyridine-4-boronic acid pinacol ester
Schmelzpunkt 97°C to 99°C
Menge 250 mg
Formelmasse 230.07
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'