missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-1-Butanol 99 %, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Menge:
100 mL
1 L
Verpackung:
Glasflasche
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemischer Name oder Material | 3-Methoxy-1-butanol |
Namenshinweis | 99% |
CAS Min. % | 98.5 |
CAS Max. % | 100.0 |
Schmelzpunkt | -85.0°C |
Farbe | Farblos |
Dichte | 0.9200g/mL |
Siedepunkt | 161.0°C |
Flammpunkt | 46°C |
Infrarotspektrum | Echt |
Show More |
Safety and Handling
Product Identifier
- 3-Methoxy-1-butanol
- Achtung
- Entzündbare Flüssigkeit Kategorie 3
- H226-Flüssigkeit und Dampf entzündbar.
- P210-Von Hitze, heißen Oberflächen, Funken, offenen Flammen und anderen ndquellen fernhalten. Nicht rauchen.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'