missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)butyraldehyd, 95 %, Thermo Scientific Chemicals

Catalog Number 11469517
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16630-52-7
C5H10OS
118.19
MFCD00036622
NCBDFIPMWRKPDU-UHFFFAOYNA-N
3-methylthio butanal, 3-methylthio butyraldehyde, butanal, 3-methylthio, 3-methylsulfanyl butanal, beta-methylthio butyraldehyde, fema no. 3374, butyraldehyde, 3-methylthio, potato butyraldehyde, 3-methylthiobutyraldehyde, butanal,3-methylthio
61845
3-Methylsulfanylbutanal
CSC(C)CC=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16630-52-7
C5H10OS
118.19
MFCD00036622
NCBDFIPMWRKPDU-UHFFFAOYNA-N
3-methylthio butanal, 3-methylthio butyraldehyde, butanal, 3-methylthio, 3-methylsulfanyl butanal, beta-methylthio butyraldehyde, fema no. 3374, butyraldehyde, 3-methylthio, potato butyraldehyde, 3-methylthiobutyraldehyde, butanal,3-methylthio
61845
3-Methylsulfanylbutanal
CSC(C)CC=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Methylthio)butyraldehyde
Siedepunkt 63°C to 65°C (11 mmHg)
Flammpunkt 62°C (144°F)
Menge 10 g
Empfindlichkeit Air Sensitive
Formelmasse 118.2
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'