missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzamidoxim, 97 %, Thermo Scientific Chemicals

Catalog Number p-7034526
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5023-94-9
C7H7N3O3
181.15
MFCD00053609
ZAIHFKLUPWFUGH-UHFFFAOYSA-N
3-nitrobenzamidoxime, n-hydroxy-3-nitro-benzamidine, m-nitrobenzamide oxime, z-n'-hydroxy-3-nitrobenzimidamide, m-nitrobenzamidoxime, n-hydroxy-3-nitro-benzenecarboximidamide, n'-hydroxy-3-nitrobenzene-1-carboximidamide, benzenecarboximidamide, n-hydroxy-3-nitro, z-n'-hydroxy-3-nitrobenzene-1-carboximidamide
6424871
(Z)-N'-hydroxy-3-nitrobenzene-1-carboximidamide
N\C(=N/O)C1=CC=CC(=C1)[N+]([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5023-94-9
C7H7N3O3
181.15
MFCD00053609
ZAIHFKLUPWFUGH-UHFFFAOYSA-N
3-nitrobenzamidoxime, n-hydroxy-3-nitro-benzamidine, m-nitrobenzamide oxime, z-n'-hydroxy-3-nitrobenzimidamide, m-nitrobenzamidoxime, n-hydroxy-3-nitro-benzenecarboximidamide, n'-hydroxy-3-nitrobenzene-1-carboximidamide, benzenecarboximidamide, n-hydroxy-3-nitro, z-n'-hydroxy-3-nitrobenzene-1-carboximidamide
6424871
(Z)-N'-hydroxy-3-nitrobenzene-1-carboximidamide
N\C(=N/O)C1=CC=CC(=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Nitrobenzamidoxime
Menge 1 g
Formelmasse 181.15
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'