Learn More
ibidi 3 Well Chamber, Removable
Artikelnummer. 15683424
Encompass_Preferred
Ansicht ändern
Click to view available options
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15683424 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15683424 Lieferant ibidi Lieferanten-Nr. 80381
Please to purchase this item. Need a web account? Register with us today!
Chamber slide with a removable 3 well silicone chamber for cell culture and immunofluorescence staining
  • Chamber slide with a removable 3 well silicone chamber
  • For cell culture and immunofluorescence staining
  • For use with upright and inverted microscopes
  • Bottom material: microscopy glass
  • Sterilized
Spezifikation
Anzahl pro Packung 15